Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Allis Tissue Forceps, Sklar
Supplier: Sklar
36-2160 36-2275 36-2260
82030-180EA 109.14 USD InStock
82030-180 82030-184 82030-182
Allis Tissue Forceps, Sklar
Forceps
Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue
Censitrac Ready OR Grade Allis Tissue Forceps are industry standard and constructed of stainless steel.
They are commonly used with tonsil, vaginal, breast, and thyroid tissue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty