Would you like to log in again?
Wash Bottles, Wide Neck, Printed, Dynalon
Supplier: Dynalon
506965-0003 506965-0002
30620-208CS 76.01 USD InStock
30620-208 30620-206
Wash Bottles, Wide Neck, Printed, Dynalon
Bottles Wash Bottles
High-quality wash bottles are manufactured from flexible natural polyethylene and fitted with white, leak-proof, wide neck polypropylene spout closures.
  • All vessels feature S.P.I. recycling codes on the base
  • Maximum operating temperature of LDPE is 80 °C (176 °F)
The wide neck offers easy spill-free filling, and the acute angled, precision-molded dispensing spout ensures a consistent downward pointing jet.
Delivery information: The wash bottles are supplied in shrink-wrapped packs of five.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty