Would you like to log in again?
SP Bel-Art Dry-Keeper™ Vertical Auto-Desiccator Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art
H42056-1003
24983-455EA 3147.2 USD InStock
24983-455
SP Bel-Art Dry-Keeper™ Vertical Auto-Desiccator Cabinet, Bel-Art Products, a part of SP
Desiccators Desiccator Cabinets
The 120VAC (11 watt) electronic dehumidification system maintains relative humidity levels at 30 to 40% (cabinet interior rH% levels can differ depending upon room temperature, room rH% and usage conditions) and keeps dry air circulating throughout the cabinet.
  • Durable
  • Clear PVC panels
  • Sleek black ABS accents
  • Vertical profile
  • Movable shelves for adjustable dry storage configurations
Permanent desiccant is contained in the control unit and a red light indicates operation of the 4½ hour absorption period while a green light indicates operation of the ½ hour regeneration of the desiccant.
The cabinet features a spacious 2 cu.ft. interior volume and a large, in-door dial hygrometer for easy visualization of rH%. Three adjustable, removable, and perforated shelves facilitate full circulation of air. Shelf rails can be repositioned on any one of 15 steps set at 28 mm intervals. The gasketed door opens to the right and closes with two latches for a tight seal. Interior opening measures 25 x 43 cm (10 x 17").
Certifications: CE Marked.
Ordering information: Additional shelves and rails sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty