Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vikan® Pipe Brush, Remco Products
Supplier: Remco
Vikan®
70355
76007-928EA 50.29 USD InStock
76007-928
Vikan® Pipe Brush, Remco Products
Brushes
The soft bristles on this brush make it useful to clean areas like small tanks and containers.
  • Often used to clean mixers for blending ingredient
  • Fits well into meat mincer barrels
  • Dishwasher safe
  • Autoclavable up to 250 °F
Block material is polypropylene; bristle material is polyester.
Requires a European-thread handle, and may be attached to any length handle to fit the user’s need. 
Certifications: FDA compliant raw material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty