Would you like to log in again?
STAR™ DISH Petri Dishes, Semi-Nesting Alpine, Phoenix
Supplier: Phoenix
370 371
25389-030CS 295.09 USD InStock
25389-030 25389-032
STAR™ DISH Petri Dishes, Semi-Nesting Alpine, Phoenix
Culture Dishes Petri Dishes
These petri dishes are most suitable for use with high-speed media pouring machines
The recessed lid has a smooth beveled angle allowing the dish to slide easily during automated pouring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty