Would you like to log in again?
STAR™ DISH Aqua Plate' Petri Dishes, Phoenix Biomedical'
Supplier: Phoenix
352
25389-020CS 367.19 USD InStock
25389-020
STAR™ DISH Aqua Plate' Petri Dishes, Phoenix Biomedical'
Culture Dishes Petri Dishes
These Aqua Plate petri dishes are ideal for membrane filtration techniques
These dishes are best when used for field sample collection. The tight-fitting lids on these dishes can be sealed with tape.
They accommodate a 47 mm round membrane filter and can be used for a variety of water testing applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty