Would you like to log in again?
STAR™ DISH ALPINE Petri Dishes, Two Sections, Phoenix
Supplier: Phoenix
372
25389-034CS 311 USD InStock
25389-034
STAR™ DISH ALPINE Petri Dishes, Two Sections, Phoenix
Culture Dishes Petri Dishes
These petri dishes are ideal when a larger surface is required for culture or antibiotic sensitivity discs
These dishes are ideal for simultaneous differential studies of organisms or media in a common environment. Costs are reduced by minimizing the use of dishes, media and space.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty