Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethylene glycol ≥99.0%, Reagent Grade

Supplier: VWR
Warning
Synonyms: Digol2,2'-Oxybisethanol2,2'-Oxybis(ethan-1-ol)

VWRV0194-1L VWRV0194-500ML
89399-814EA 75.67 USD InStock
89399-814 89399-812
Diethylene glycol ≥99.0%, Reagent Grade
Diethylene glycol


Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.118 g/cm³ (20 °C)
Flash Pt: 123 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/97/2297797.jpg
Order Now
Specification Test Results
Color (APHA) = 10
Purity = 99 %
Refractive Index 1.445 - 1.450

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty