Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)

Supplier: VWR
Synonyms: DNA sodium saltThymonucleic acid sodium salt

VWRV1B1509-25G
89422-824EA 126.17 USD InStock
89422-824
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
DNA sodium salt (Deoxyribonucleic acid sodium salt)


MDL Number: MFCD00130921
CAS Number: 9007-49-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty