Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chlorite, high purity

Supplier: VWR
Danger

VWRVN819-100G
97066-116CS 856.75 USD InStock
97066-116
Sodium chlorite, high purity
Sodium chlorite


Formula: ClNaO₂
MW: 90.44 g/mol
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: 250…257 °C
Density: 1.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1496
ADR: 5.1,II
Merck Index: 12,08743
Order Now
Specification Test Results
Arsenic 3 ppm
Heavy Metals (as Pb) 20 ppm
Hydrogen Peroxide 0.01 %
Lead 10 ppm
Mercury 0.1 ppm
Sodium Carbonate 2 %
Sodium Chlorate 4 %
Sodium Chloride 11 % - 19 %
Sodium Chlorite 77.5 % - 82.5 %
Sodium Hydroxide 3 %
Sodium Sulfate 3 %
Water (by difference) 6 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty