Would you like to log in again?
ESD Shelf Bins
Supplier: Treston
SBS6-3010-4ESD SBS6-3020-4ESD
76015-070PK 108.82 USD InStock
76015-070 76015-062
ESD Shelf Bins
Boxes ESD-Safe Boxes
These black shelf bins are designed to fit all standard metric shelves, cabinets, paternosters and industrial workstations. Made from semiconductive polypropylene (PP).
  • Bins of the same width stack securely, even if they are of different depths
  • Straight sides ensure an efficient use of space
  • Withstands the effects of oils, most acids and solvents
  • Suitable for use in cleanroom environments
  • Four depth options: 11.81, 15.75, 19.69 or 23.62"
  • Corrugated base for ease of picking up items
  • Multiple colors available
The shelf bins are particularly suitable for use in small parts retrieval systems and mobile shelving. Corrugated bases ease the picking of very small items. Full width label with protective shield is easy to keep clean and change when necessary.
Ordering information: Labels included. Dividers sold separately. Installation is required. Assembly videos available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty