Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
GeneMate Mini-V Mixer, Benchmark Scientific
Supplier: Benchmark Scientific
BV101-P-GM
490015-280EA 299.85 USD InStock
490015-280
GeneMate Mini-V Mixer, Benchmark Scientific
Shakers and Mixers Vortex Mixers
The Mini-V Mixer instantly vortexes even the largest samples, including nearly full 50 mL tubes
Ordering Information: The Mini-V Mixer comes with a two year, full-replacement warranty.
Insta-Touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth, quiet and efficient. When presure is removed, operation immediately ceases. The unique head design prevents liquids from entering the housing, prolonging motor life.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty