Would you like to log in again?
PTFE Tube Compression to Male NPT, Bore-Through, Ace Glass Incorporated
Supplier: Ace Glass
12710-02 12710-30 12710-14 12710-32 12710-08 12710-04 12710-06 12710-40 12710-36 12710-34 12710-10 12710-42 12710-38 12710-12
10099-522EA 112.08 USD InStock
10099-522 10099-302 10099-312 10099-304 10099-314 10099-306 10099-316 10099-308 10099-516 10099-298 10099-520 10099-300 10099-310 10099-518
PTFE Tube Compression to Male NPT, Bore-Through, Ace Glass Incorporated
Fittings and Connectors
PTFE tube compression fitting to male NPT with tubing O.D bored through body.
Body and ferrules made of virgin PTFE. The nut and ferrule-gripper is made of PVDF. Performance at ambient room conditions is 120psi for 1/16" fittings linear decreasing to 80psi for 3/4" fittings and 60psi for fittings larger than 3/4". Performance at elevated temperatures up to 85°C/185°F: 90psi for 1/16" fittings linear decreasing to 60psi for 3/4" fittings and 40psi for fittings larger than 3/4".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty