Would you like to log in again?
PTFE Tube Compression Elbow Union, Ace Glass Incorporated
Supplier: Ace Glass
12716-10 12716-14 12716-02 12716-16 12716-12 12716-04 12716-08
10099-368EA 79.61 USD InStock
10099-368 10099-378 10099-380 10099-370 10099-372 10099-374 10099-376
PTFE Tube Compression Elbow Union, Ace Glass Incorporated
Fittings and Connectors
PTFE Tube Compression Elbow Union
Body and ferrules made of virgin PTFE. The nut and ferrule-gripper is made of PVDF. Performance at ambient room conditions: 120 PSI for 1/16" fittings linear decreasing to 80 PSI for 3/4" fittings and 60 PSI for fittings larger than 3/4". Performance at elevated temperatures up to 85C/185F: 90 PSI for 1/16" fittings linear decreasing to 60 PSI for 3/4" fittings and 40 PSI for fittings larger than 3/4".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty