Would you like to log in again?
Vernier® Melt Station
Supplier: Vernier
MLT-TUBE MLT-BTA
470219-304EA 23.99 USD InStock
470219-304 470113-968
Vernier® Melt Station
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Accurately determine the melting temperature of a solid.
  • Accurate results using built-in RTD over a range of 30°C to 260°C
  • Accuracy ± 0.31 + 0.0006T, where T is the temperature in Celsius
  • Resolution 0.10°C
  • High-quality 6X viewing lens for clear observation of the samples
  • Innovative, adjustable tilt allows for optimal viewing angle
  • Cooling fan reduces time between tests
Red LED (indicates heating mode)
Blue LED (indicates cooling mode with cooling fan running)
Yellow LED (safety shut off activated)
Connect the Melt Station to a Vernier interface to measure the melting temperature of solid substances.
For educational use only.
Ordering information: Each Melt Station comes with 100 standard capillary tubes. Additional packages of 100 are available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty