Would you like to log in again?
Tissue Primer, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K054
10064-762EA 252.62 USD InStock
10064-762
Tissue Primer, Diagnostic BioSystems
Assays Immunological Assays
Tissue Primer is a universal enzyme blocking reagent to use in horse radish peroxidase (HRP) and alkaline phosphatase (AP) based immunohistochemical staining procedures done on paraffin tissue sections, frozen tissue sections and cell smears to suppress nonspecific staining due to endogenous enzyme activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty