Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Goniometer

Supplier: VERNIER SOFTWARE & TECH SE
GNM-BTA
470112-950EA 188.99 USD InStock
470112-950
Goniometer
Goniometers
The Goniometer measures the angle of a joint, such as the knee or elbow. Use it to measure the range of motion of a limb during different types of physical activity.

  • Lightweight and flexible joint arms allow the limb to move naturally
  • Requires a compatible interface and software to collect data

A set of elastic straps can be used to secure the sensor to the subject for dynamic measurements of joint angle during different types of physical activity. You can also use it with a Vernier EKG Sensor to measure muscle activity during different phases of limb motion. In addition, the sensor head can be detached from both arms of the sensor for use in STEM and engineering activities.

Range: 0 - 340° (or ±170°)
Accuracy: ±3° / ±1° (with calibration)
12-bit resolution: 0.12°
Stored Calibration Values: 90°/V (slope); –45° (intercept)

Ordering information: Interface and software sold separately.

Delivery information: Goniometer includes joint angle sensor with base plate attached to static arm; 12-inch flexible joint arm; 10-inch flexible joint arm; 8-inch flexible joint arm; Two 24-inch elastic, hook-and-loop straps; Two 14-inch elastic, hook-and-loop straps; User manual.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty