Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Potassium Ion-Selective Electrode

Supplier: VERNIER SOFTWARE & TECH SE
K-BTA
470112-984EA 298.99 USD InStock
470112-984
Vernier® Potassium Ion-Selective Electrode
Electrodes ISE Electrodes
Measure the concentration of potassium ions in aqueous solutions.

  • Stick-style sensor

The Potassium Ion-Selective Electrode makes the measure of potassium ions portable. When used with a Vernier LabQuest 2, the Potassium ISE can be used to take readings in the field – at lakes or streams. This sensor is easier to set up and use than other methods of measuring potassium ion concentration in aqueous solutions. Because it is a stick-style sensor, its use is similar to using a conductivity probe or a pH sensor in a laboratory setting or in the field.

Ion-Selective Electrodes require good chemical technique and careful calibration to obtain accurate results. They are not recommended for middle school or elementary students.

We recommend that you do not purchase Potassium Ion-Selective Electrode replacement modules too far in advance of their expected time of use; degradation occurs while replacement modules are stored on the shelf.

Interfering Ions: Rb2+, Cs2+, NH4+, Ca2+, Mg2+, Li+
Electrode Resistance: >50 mΩ
Electrode Slope: 56 +/- 4 mV/decade at 25°C
Calibration Voltage (typical): 2.7 V (1000 ppm), 1.9 V (10 ppm)
Reproducibility (precision): +/- 11% of full scale
Minimum sample size: 3 mL in a 50 mL beaker

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty