Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sarcosine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: N-MethylglycineH-Sar-OHMethylaminoacetic acid

M-Clarity™ quality level = MQ200

8.07666.9010 8.07666.9010
EM8.07666.9010EA 3192 USD InStock
EM8.07666.9010 CA8.07666.9010
Sarcosine for synthesis, Sigma-Aldrich®
Sarcosine


Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00004279
CAS Number: 107-97-1
Order Now
Specification Test Results
Solubility 1481 g/l (20°C)
Melting Point 208 °C
Molar Mass 600 kg/m3
Bulk Density 600 kg/m3
pH 6.1 (100 g/l, H2O, 20 °C)
Synonyms Methylaminoacetic acid, N-Methylglycine
Hill Formula C3H7NO2
Chemical Formula CH3NHCH2COOH

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty