Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD19 Mouse Monoclonal Antibody (violetFluor® 450) [clone: HIB19]

Supplier: Tonbo Biosciences
75-0199-T100 75-0199-T025
10051-800EA 96.5 USD InStock
10051-800 10051-802
Anti-CD19 Mouse Monoclonal Antibody (violetFluor® 450) [clone: HIB19]
Antibodies
The HIB19 antibody reacts with human CD19, a 95 kDa glycoprotein which acts as a co-receptor, along with CD21, CD81 and CD225, in support of the functional B cell receptor (BCR). This complex provides antigen-specific recognition and subsequent activation of B cells to proliferate and differentiate into antibody-secreting cells (plasma cells) or memory B cells, which are crucial for secondary antigen encounter. CD19 is a lineage-differentiation marker, as its expression is detectable at the earliest B cell stages, through development, and is finally lost upon transition to mature plasma cells.

The HIB19 antibody is widely used as a phenotypic marker for CD19 expression on B cells, as well as on dendritic cell subsets.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty