Would you like to log in again?
Nalgene™ Griffin Low-Form Beaker Variety Pack, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
1201-1234
10124-798CS 76.75 USD InStock
10124-798
Nalgene™ Griffin Low-Form Beaker Variety Pack, Thermo Scientific
Beakers
A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab.
  • No drip single-spout design provides precision pouring control
  • With ribs for easy stacking
  • Impact resistant at temperatures above −40 °C
  • Autoclavable/graduated compliance
Excellent chemical resistance to a wide variety of lab reagents, easy to read silk screened graduations and convenient stacking capabilities make these beakers an excellent choice for daily lab activities.
Silk screened with graduations, size code, resin code, catalog number, max/temperature and 'no flame' symbol for easy identification and safe use. Meets ISO/DIN 7056 International Standards for laboratory plasticware beakers and 21 CFR Part 177.1520 for food/beverage use.
Certifications: Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Beakers and 21 CFR Part 177.1520 for food/beverage use.
Ordering information: Thermo Scientific™ Nalgene™ Griffin Low-Form Beaker Variety Pack includes one each 50, 100, 150, 250, 400, 600 and 1000 ml PPCO beakers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty