Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nunc® Round-Bottom Tubes, 14mL, Thermo Scientific

Supplier: Thermo Scientific
Nunc®
150268 150268
89134-938CS 347.13 USD InStock
CA89134-938 89134-938
Nunc® Round-Bottom Tubes, 14mL, Thermo Scientific
Tubes Culture Tubes
Perform critical cellular applications with the Nunc® 14 ml round-bottom tubes.

Optimized gas exchange with dual position vent/close cap. Off-gas volatile organic compounds easily with breathable packaging. Certificates of conformity available for every lot. Designed for research use only.

Certifications: Mouse Embryo Assay (MEA) tested. Produced under ISO 13485 with full lot traceability. USP Class VI tested. Certified.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty