Would you like to log in again?
Somso® Artificial Human Skeleton Male
Supplier: MARCUS SOMMER
Somso®
QS 10/6+9
470105-194EA 3995.99 USD InStock
470105-194
Somso® Artificial Human Skeleton Male
Models Human Biology Models
Somso® premium quality models.
  • Five year warranty
Natural cast of a male adult skeleton made of SOMSO-Plast®. On the right side of the body the bones are numbered and the ligaments of the shoulder, elbow, hip and knee are shown for anatomically accurate demonstration of joint movement. The points of origin and attachment of the most important muscles from head to foot are marked in colour on the left side of the body (origin red, attachment blue). Mounted on a stand with rollers.
Ordering information: Items on this page are not stocked and may have an extended time for delivery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty