Would you like to log in again?
Vertebrate Brains, ED Speldy East Co. Ltd.
Supplier: ED SPELDY EAST CO. LTD.
T603-B
470150-804EA 141.65 USD InStock
470150-804
Vertebrate Brains, ED Speldy East Co. Ltd.
Specimens Preserved Specimens Display Specimens
This kit looks at the similarities and differences between vertebrate brains
There is one specimen block included (6.5x3.0x1.0 in.). Specimens are preserved and encased in clear resin material. They can be observed from every side to see details. Specimen include: fish (Carrassias auratus), amphibian (Rana limnocharis), reptile (Enhydris chinensis), bird (Parus major), and mammal (Lepus sinensis). Teacher information guide, student activity sheet, and answer key are included. Size of product is 8.4x4.6x1.5in.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty