Would you like to log in again?
Microwave Leakage Detector, Extech
Supplier: Extech
EMF300
10068-324EA 357.66 USD InStock
10068-324
Microwave Leakage Detector, Extech
Radiation Meters
Easy-to-use, compact microwave leakage detector designed to measure high frequency radiation levels emitted from household and commercial microwave ovens.
Unit features an audible alarm and overrange indicator.
Certifications: CE
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty