Would you like to log in again?
Compact Hygro-Thermo-Anemometer, Extech
Supplier: Extech
EN100
10068-412EA 562.44 USD InStock
10068-412
Compact Hygro-Thermo-Anemometer, Extech
Anemometers
11-in-1 Environmental Meter with built-in sensors to measure air velocity, air flow, temperature, humidity, wet bulb, dew point, heat index, wind chill, barometric pressure, altitude and light.
Field replaceable, low friction vane wheel improves air velocity accuracy. Backlit LCD display features a data hold function and an auto-off after 5 seconds. Display rotates, depending on function in use.
Certifications: CE
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty