Would you like to log in again?
Fluoride ISE Standards, Ricca Chemical
Supplier: RICCA Chemical Company
3161.5-32
RC3161.5-32EA 104.2 USD InStock
RC3161.5-32
Fluoride ISE Standards, Ricca Chemical
Standards Anionic Standards Fluoride Standards
Fluoride standard, 1 mL = 0.0005 mg F-, 0.5 ppm F-, premixed with TISAB.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty