Would you like to log in again?
KWIK-STIK Citrobacter koseri ATCC® 27156™, Microbiologics
Supplier: Microbiologics
KWIK-STIK™
0106P 0106K
89502-094EA 321.1 USD InStock
89502-094 89502-098
KWIK-STIK Citrobacter koseri ATCC® 27156™, Microbiologics
Control reagents
The KWIK-STIK™ is well-known for its clever design and accurate, repeatable results
Each KWIK-STIK™ device features a single QC microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab. For added convenience, a peel-off identification label is included for easy documentation.
Certifications: The KWIK-STIK™ falls under the category of In-Vitro Diagnostics Medical Devices Directives for CE Mark Conformity. This product is also listed with the FDA.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the vwr.com product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty