Would you like to log in again?
Whole Rabbit Sera, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
B309 B309
RLB309EA 249 USD InStock
CAB309 RLB309
Whole Rabbit Sera, Rockland Immunochemicals
Sera (Serum)
The whole sera are prepared from normal sera by a multi-step process that includes delipidation and selective precipitation
All sera are derived from starting materials of North American origin and collection was performed in USDA inspected facilities. All materials have been inspected and certified to be free of disease and are suitable for exportation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty