Would you like to log in again?
Whole Rat Sera, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
B310 B310
RLB310EA 518 USD InStock
RLB310 CAB310
Whole Rat Sera, Rockland Immunochemicals
Sera (Serum)
The whole sera are prepared from normal sera by a multi-step process that includes delipidation and selective precipitation
All sera are derived from starting materials of North American origin and collection was performed in USDA inspected facilities. All materials have been inspected and certified to be free of disease and are suitable for exportation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty