Would you like to log in again?
Sterile Goat Sera, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
D104-00-0050 D104-00-0050
RLD104-00-0050EA 373.85 USD InStock
CAD104-00-0050 RLD104-00-0050
Sterile Goat Sera, Rockland Immunochemicals
Sera (Serum)
Sera is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: For heat-inactivated sera, contact your Avantor sales representative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty