Would you like to log in again?
Ward's® 4-Way Tube Racks
Supplier: Heathrow Scientific
Ward's®
HS29022GV
470199-488CS 158.95 USD InStock
470199-488
Ward's® 4-Way Tube Racks
Racks Tube Racks
Link racks together to create larger racks as needed.
  • Made polypropylene that is chemical resistant
  • Easy to clean
  • Autoclavable at 121 °C
Used as standalone unit or infinite units can be linked to create configurations to fit number of wells and tube sizes needed. Molded alphanumeric identification for quick location and retrieval of tubes. Racks can be used in water baths and placed in freezer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty