Would you like to log in again?
Worksurface Add-on, Production Basics
Supplier: Production Basics
8982 PB-8982
87000-386EA 93.65 USD InStock
87000-386 76243-686
Worksurface Add-on, Production Basics
Furniture Casework
Add any of these unique laminate worksurface features when ordering a laminate worksurface or workstation.
  • Ergo add-on brings the task closer to the operator with the Ergo worksurface
  • Works only with C-leg or easy-lift dual leg laboratory benches
  • Reduces an operator’s reach area
  • Bullnose front edge add-on adds a 180° laminated radius edge to any worksurface for a safe and comfortable work area
This unique laminate worksurface features a angled cut-out in the front center of the station measuring 91.4 cm (36") wide at front, 15.2 cm (6") deep indentation and is 61 cm (24") wide at back angle. Finished with laminate on the front and PVC on sides and back in a matching color.
Compliments production basics lab bench worksurface.
Ordering information: Must purchase with a lab bench or worksurface, sold separately, for installation. Purchase lab bench or worksurface and add-on desired part number option.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty