Would you like to log in again?
Security-Snap™ Sterile Coliform Water Sample Bottle, Thermo Scientific
Supplier: Thermo Scientific
Security-Snap™
411G10R27
73520-380CS 430.86 USD InStock
73520-380
Security-Snap™ Sterile Coliform Water Sample Bottle, Thermo Scientific
Bottles Water Sampling Bottles
The Security-Snap™ Sterile Coliform Water Sample Bottle meets all the requirements for microbiology sample containers as specified by the 'EPA Manual for the Certification of Laboratories Analyzing Drinking Water'.
Manufactured from non-toxic, non-autofluorescent polypropylene with a molded-in 100±2.5 ml fill line.
The bottle features a Security-Snap™ flip-top lid which provides tamper evidence before and after sample collection, while protecting against any possibility of accidental opening during transport.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty