Would you like to log in again?
Custody Seals, Thermo Scientific
Supplier: Thermo Scientific
500
IR500PK 87.19 USD InStock
IR500
Custody Seals, Thermo Scientific
Seals Security Seals
Custody Seals provide complete chain-of-custody and tamper evidence for samples from the point of collection to lab receipt.
Custody seal adhesive stickers securely seal sample bottles and cannot be peeled off. The only way to open the sealed bottle is to break the paper label. Each seal includes a writing area for custody signature and date for tamper-evident secutity. Apply at the point of sample collection, and ship samples to their destination with confidence.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty