Would you like to log in again?
Screw Cap Micro Tubes, Type H, Sarstedt
Supplier: Sarstedt
72.608 72.693 72.693.005 72.693.100
101093-787PK 121.23 USD InStock
101093-787 101093-865 101093-579 101093-750
Screw Cap Micro Tubes, Type H, Sarstedt
Tubes Microtubes
Type H micro tubes are designed for transport, storage, and sample processing in the laboratory.
All tubes feature a non-skirted base. Tubes and caps are manufactured from polypropylene, and are autoclavable at 121 °C.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty