Would you like to log in again?
Inoculating Loops, Nickel-Chrome, Non-Calibrated, COPAN Diagnostics
Supplier: COPAN Diagnostics
CAS311-25
30002-134PK 250.04 USD InStock
30002-134
Inoculating Loops, Nickel-Chrome, Non-Calibrated, COPAN Diagnostics
Spreaders Inoculating Loops and Needles
Non-calibrated loops are available in a range of sizes with internal diameters (Ø) of 3, 4 or 5mm
Loops are made of an 80/20 Nickel-Chrome metal wire. A strand of wire is molded around a specific pin size to form the loop shape and then the ends of wire strand are twisted together. The twisting process gives the loop its characteristic flexibility.
Ordering information: Loops are supplied in tubes containing 25 loops.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty