Would you like to log in again?
SecureSeal™ Thermal Adhesive Sealing Film, Simport Scientific
Supplier: Simport
T329-6
10030-948PK 348.88 USD InStock
10030-948
SecureSeal™ Thermal Adhesive Sealing Film, Simport Scientific
Seals Microplate Seals
SecureSeal™ Thermal Adhesive Sealing Film is designed for real time qPCR application and features a special pressure sensitive DMSO-resistant silicone adhesive embedded inside the film itself.
A polyester backing with end tabs enables easy positioning on the plate. This polyolefin film is also perfectly suited for use with raised rim plates, ensuring reliable sealing around each well. SecureSeal™ Thermal Adhesive Sealing Film offers the highest transparency, and has an operating temperature range of –40 to 120°C (–40 to 248°F). Certified RNase-, DNase-, and DNA-free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty