Would you like to log in again?
KIMBLE® Right Angle Flow Control Inlet Adapter with Varibor Metering Plug, DWK Life Sciences
Supplier: DWK Life Sciences
185031-2440
KT185031-2440EA 405.66 USD InStock
KT185031-2440
KIMBLE® Right Angle Flow Control Inlet Adapter with Varibor Metering Plug, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Allows delivery of preset volumes of gases or liquids to reactions using the needle valve.
  • Provides on/off control with main valve
  • Inner [ST] joint on one end
  • 90° angle hose connection on opposite end
Adapter provides on/off control with the main valve
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Features an inner Standard Taper joint on one end and 90° hose connection on the opposite end.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty