Would you like to log in again?
Buret Filler, Eisco Scientific
Supplier: Eisco Scientific
BTAUT50
470121-260EA 71.69 USD InStock
470121-260
Buret Filler, Eisco Scientific
Burets
Automatic 50ml burette made of borosilicate glass which is self-zeroing
0.1ml resolution with permanent blue markings against a white background for easy viewing.  Stopcock is pushbutton style for ease of use to deliver fluids drop by drop.   Attached to the stop cock is a screw clamp for repeat titrations.  This set up is a complete self-supporting structure with a heavy duty base made of polyethylene which the reservoir snaps into.  The advantage to a system such as this is that it is a completely closed system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty