Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Whatman™ Grade GF 92 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10421030 10421026 10421043 10421030 10421060 10421026 10421060 10421043 10421019 10421019 10421057 10421057
10035-012PK 241.93 USD InStock
CA11013-248 10035-012 CA11013-256 10035-010 CA11012-918 CA11013-254 10035-016 10035-014 CA11013-052 CA11013-250 10035-008 10035-018
Whatman™ Grade GF 92 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
Whatman Grade GF 92 glass microfiber filters from Cytiva are designed for prefiltration in order to protect against membrane fouling. Grade GF 92 is strengthened with an inorganic binder.
  • Inorganic binder provides increased strength
  • Temperature resistant up to 500 °C
  • Medium filtration speed
Cytiva's business offers a variety of binder-strengthened glass microfiber filters for your filtration and prefiltration needs, including environmental, air pollution, water analysis.
Prefiltration can improve performance and longevity of equipment in applications such as:
- Measurement of crop protection agent using gas chromatography (GC) or HPLC
- Separation and determination of cold sludge when brewing beer
- Soot separation prior to use of gas analyzers
- Roll filters for automatic air filtration units
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty