Would you like to log in again?
3B Scientific® Shoulder Joint With Rotator Cuff
Supplier: AMERICAN 3B SCIENTIFIC SE
1000176
470103-190EA 429.25 USD InStock
470103-190
3B Scientific® Shoulder Joint With Rotator Cuff
Models Human Biology Models
This shoulder joint model consists of the upper half of the humerus, as well as the clavicle and scapula.
  • Show the structure of the knee in sections
  • 3 year manufacturers warranty
  • Includes 3B smart anatomy
Besides showing the musculature of the rotator cuff, the shoulder joint model also shows the origin and insertion points of the shoulder muscles highlighted in color (origin = red, insertion points = blue). Upon detaching the individual muscles, all movements of the shoulder joint can be carried out.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty