Would you like to log in again?
Whatman™ Application-Specific Filter Papers, Grade 3000, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10316114 10316114
10034-740PK 174.13 USD InStock
CA11012-686 10034-740
Whatman™ Application-Specific Filter Papers, Grade 3000, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Smooth filter papers with good retentivity at a relatively high filtration speed. They are used for the clarifying filtration of dried beet pulp extracts or beet juice after the addition of lead acetate for subsequent polarimetric sugar determination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty