Would you like to log in again?
Whatman™ MV050 Series Vacuum Filtration Apparatus, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10440020 10440000 10440020 10440000
10035-020PK 1501 USD InStock
CA11012-926 10035-020 10035-026 CA11013-082
Whatman™ MV050 Series Vacuum Filtration Apparatus, Whatman products (Cytiva)
Flasks Filter Flasks
MV series vacuum filtration devices are made of stainless steel, which is especially suitable for microbiological applications.
The system can be used up to 200ºC, is autoclavable, and can be sterilized by dry heat up to 180ºC.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty