Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Polydisc HD In-Line Filters, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Whatman™ Polydisc™ Polydisc™ Whatman™
2228 6728-5050 6728-5100 6728-5100 2228 2227 6728-5050 2227
10035-760PK 635.55 USD InStock
10035-760 10035-758 10147-996 CA11013-066 28137-747 CA28137-747 CA10147-996 CA11012-820
Whatman™ Polydisc HD In-Line Filters, Whatman products (Cytiva)
Filters In-Line Filters
Polydisc HD (Heavy Duty) is a durable all-polypropylene in-line filter for large volumes of liquid samples containing even harsh chemical solvents.

  • In-line, hose-connected filter with up to 1000 ml capacity
  • High flow filter with durable all-polypropylene construction
  • Broad aqueous and solvent compatibility
  • 99% liquid retention rating
  • Available in 5 and 10 µm pore size
  • Recyclable after use

Labs that regularly process large sample volumes with different chemical compositions, pH values, and temperatures require a filter that is fast, durable, and broadly compatible.

Heavy-duty Polydisc filter is a durable all-polypropylene filter. Its high strength, aqueous and solvent compatibility, and flow rate make it an excellent in-line filter for up to 1000 ml sample volumes

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty