Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

n-Hexane, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: HexanenHexaneHexyl hydride

M-Clarity™ quality level = MQ200

1.04368.9011 1.04368.9190 1.04368.2511 1.04368.6025 1.04368.2500 1.04368.2500 1.04368.6025 1.04368.1000 1.04368.9011 1.04368.2511 1.04368.9190
EM1.04368.9190EA 4291 USD InStock
EM1.04368.9190 CA1.04368.9190 EM1.04368.2511 EM1.04368.2500 CA1.04368.6025 EM1.04368.9011 CA1.04368.2500 CA1.04368.2511 CA1.04368.9011 EM1.04368.6025 EM1.04368.1000
n-Hexane, EMPLURA®, Supelco®
n-Hexane


Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty