Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium carbonate ≥98.5% (by acidimetry), Supelco®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ200

1.05670.9050 1.05670.9050
EM1.05670.9050EA 15669 USD InStock
EM1.05670.9050 CA1.05670.9050
Lithium carbonate ≥98.5% (by acidimetry), Supelco®
Lithium carbonate


Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Density: 2.1 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty