Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quartz, fine-granular for analysis, washed, calcinated, Supelco®

Supplier: MilliporeSigma
Synonyms: Sand, white quartzSandSilicium (IV) oxideSilicon dioxideSilicium dioxide

M-Clarity™ quality level = MQ200

1.07536.1000 1.07536.1000 1.07536.0250 1.07536.5000
EM1.07536.1000CS 3701 USD InStock
EM1.07536.1000 CA1.07536.1000 EM1.07536.5000 EM1.07536.0250
Quartz, fine-granular for analysis, washed, calcinated, Supelco®
Quartz


Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1427 °C
Density: 2.622…2.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00217788
CAS Number: 14808-60-7
EINECS: 238-878-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/83/16640583.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty