Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cadmium oxide, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Cadmium (II) oxide

M-Clarity™ quality level = MQ200

1.02015.5000 1.02015.5000
EM1.02015.5000EA 808 USD InStock
CA1.02015.5000 EM1.02015.5000
Cadmium oxide, Supelco®
Cadmium oxide


Formula: CdO
MW: 128.41 g/mol
Boiling Pt: 1559 °C (1013 hPa)
Melting Pt: ∼1426 °C
Density: 8.15 g/cm³ (20 °C)
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,I
Merck Index: 12,01659
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty