Would you like to log in again?
Reli® Quincke Needles with Metric Markings, Sterile, MYCO Medical
Supplier: MYCO Medical
SNME20G351 SNME25G351 SNME22G351 SNME22G601 SNME18G801
10148-740BX 293.57 USD InStock
10148-740 10148-714 10148-736 10148-746 10148-728
Reli® Quincke Needles with Metric Markings, Sterile, MYCO Medical
Syringe Needles
Reli Quincke Needles are high quality needles with large, clear, color-coded hubs for enhanced tactile feel and easy gauge identification
Matched-Point grinding allows for tight tolerance between the stylet and cannula to provide for smooth, low-force insertion. Reli Quincke Needles with Metric Markings have echogenic stylets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty